The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080457.3 | NP_536705.3 | 2 - 3 | 7489 | 93 |
GenBank mRNA | AF441786.1 | - | - | 7019 | 93 |
CB522483.1 | - | 1 - 2 | 373 | 93 |