Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclk1
Entrez Gene ID
13175
Gene Name
doublecortin-like kinase 1
Gene Aliases
1700113D08Rik, 2810480F11Rik, AI836758, Click-I, Cpg16, Dcamkl1, Dcl, Dclk, mKIAA0369
Chromosome Location
Chr.3: 55242369 - 55539068 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111053.1 NP_001104523.1 6 - 7 1279 84
NM_001195540.1 NP_001182469.1 2 - 3 418 84
GenBank mRNA AB073969.1 - 2 - 3 226 84
AK147831.1 - 6 - 7 1279 84
AK164993.1 - 6 - 7 1237 84
BC021354.1 - 6 - 7 1221 84
BC050903.1 - 6 - 7 1170 84
BM933369.1 - 2 - 3 411 84

View Full Product Details