Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Chtf8
- Entrez Gene ID
-
214987
- Gene Name
- CTF8, chromosome transmission fidelity factor 8
- Gene Aliases
-
5830457O10Rik, Ctf8, Derpc
- Chromosome Location
-
Chr.8: 106883863 - 106893593 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK165993.1 |
- |
1 - 2 |
97 |
98 |
View Full Product Details