The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146053.1 | NP_001139525.1 | 1 - 2 | 37 | 55 |
GenBank mRNA | BB575839.2 | - | 1 - 2 | 37 | 55 |