Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccl1
Entrez Gene ID
20290
Gene Name
chemokine (C-C motif) ligand 1
Gene Aliases
BF534335, I-309, Scya1, Tca-3
Chromosome Location
Chr.11: 82176666 - 82179884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC042744.1 - - 1613 125

View Full Product Details