The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008971.4 | NP_032997.3 | 8 - 9 | 935 | 68 |
GenBank mRNA | AK077410.1 | - | 8 - 9 | 994 | 68 |
AK147990.1 | - | 8 - 9 | 929 | 68 | |
AK150852.1 | - | 8 - 9 | 958 | 68 | |
AK165561.1 | - | 7 - 8 | 864 | 68 | |
AK168142.1 | - | 8 - 9 | 953 | 68 | |
AK168161.1 | - | 8 - 9 | 953 | 68 | |
AY267188.1 | - | 8 - 9 | 887 | 68 | |
BC015081.1 | - | 8 - 9 | 952 | 68 | |
BC094034.1 | - | 8 - 9 | 935 | 68 | |
U82324.1 | - | 8 - 9 | 943 | 68 |