Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map3k7
Entrez Gene ID
26409
Gene Name
mitogen-activated protein kinase kinase kinase 7
Gene Aliases
B430101B05, C87327, Tak1
Chromosome Location
Chr.4: 31962296 - 32023470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172688.3 NP_766276.1 11 - 12 1362 65
GenBank mRNA AK046565.1 - 11 - 12 1328 65
AK053911.1 - 11 - 12 1348 65
AK133642.1 - 5 - 6 489 65
AK137046.1 - 11 - 12 1363 65
AK159277.1 - 11 - 12 1326 65
D76446.1 - 11 - 12 1362 65

View Full Product Details