The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205108.1 | NP_001192037.1 | - | 281 | 159 |
GenBank mRNA | AY244797.1 | - | - | 282 | 159 |