Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssxb6
Entrez Gene ID
668976
Gene Name
synovial sarcoma, X member B, breakpoint 6
Gene Aliases
-
Chromosome Location
Chr.X: 8542179 - 8548362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205108.1 NP_001192037.1 - 332 151
GenBank mRNA AY244797.1 - - 333 151

View Full Product Details