Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfwd3
Entrez Gene ID
234736
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
BC027246
Chromosome Location
Chr.8: 111270944 - 111300260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146218.4 NP_666330.2 10 - 11 1797 122
GenBank mRNA AK137499.1 - 10 - 11 1777 122
AK137698.1 - 10 - 11 1760 122
AK139032.1 - 10 - 11 1779 122
BC018533.1 - 10 - 11 1766 122
BC023694.1 - 10 - 11 1802 122
BC027246.1 - 5 - 6 748 122
BC096602.1 - 10 - 11 1760 122

View Full Product Details