Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfwd3
Entrez Gene ID
234736
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
BC027246
Chromosome Location
Chr.8: 111270944 - 111300260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146218.4 NP_666330.2 2 - 3 591 89
GenBank mRNA AK137499.1 - 2 - 3 571 89
AK137698.1 - 2 - 3 554 89
AK139032.1 - 2 - 3 571 89
AK146451.1 - 2 - 3 561 89
BC018533.1 - 2 - 3 560 89
BC023694.1 - 2 - 3 596 89
BC096602.1 - 2 - 3 554 89

View Full Product Details