Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm6793
- Entrez Gene ID
-
627828
- Gene Name
- heterogeneous nuclear ribonucleoprotein A3 pseudogene
- Gene Aliases
-
EG627828
- Chromosome Location
-
Chr.8: 112013433 - 112015036 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033513.1 |
|
1 - 1 |
156 |
108 |
GenBank mRNA |
BC157904.1 |
- |
1 - 1 |
156 |
108 |
BC158074.1 |
- |
1 - 1 |
156 |
108 |
BC172011.1 |
- |
1 - 1 |
156 |
108 |
- Gene Symbol
- Hnrnpa3
- Entrez Gene ID
-
229279
- Gene Name
- heterogeneous nuclear ribonucleoprotein A3
- Gene Aliases
-
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3
- Chromosome Location
-
Chr.2: 75659230 - 75669407 on Build GRCm38
View Full Product Details