Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alkbh6
Entrez Gene ID
233065
Gene Name
alkB, alkylation repair homolog 6 (E. coli)
Gene Aliases
AI839550, Abh6, C130099A02
Chromosome Location
Chr.7: 30308753 - 30314303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198027.2 NP_932144.2 4 - 5 289 116
GenBank mRNA AK082058.1 - 4 - 5 255 116
BC029805.1 - 4 - 5 241 116
BC080310.1 - 3 - 4 210 116

View Full Product Details