The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013897.2 | NP_038925.1 | 1 - 2 | 139 | 156 |
GenBank mRNA | AA204614.1 | - | 1 - 2 | 128 | 156 |
AF196314.1 | - | - | 128 | 156 | |
AK003382.1 | - | 1 - 2 | 98 | 156 | |
AK004190.1 | - | - | 128 | 156 | |
AK012925.1 | - | 1 - 2 | 106 | 156 | |
AK018718.1 | - | 1 - 2 | 106 | 156 | |
AK019422.1 | - | 1 - 2 | 114 | 156 | |
AK169258.1 | - | 1 - 2 | 118 | 156 | |
BC029239.1 | - | 1 - 2 | 92 | 156 |