Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2a22
Entrez Gene ID
233005
Gene Name
cytochrome P450, family 2, subfamily a, polypeptide 22
Gene Aliases
EG233005
Chromosome Location
Chr.7: 26931631 - 26939412 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101467.1 NP_001094937.1 4 - 5 673 94
GenBank mRNA BC151116.1 - 4 - 5 667 94
BC158034.1 - 4 - 5 667 94

View Full Product Details