Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r9
Entrez Gene ID
435864
Gene Name
vomeronasal 2, receptor 9
Gene Aliases
EG435864
Chromosome Location
Chr.5: 108842947 - 108852510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104621.1 NP_001098091.1 2 - 3 479 131

View Full Product Details