Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r69
Entrez Gene ID
330581
Gene Name
vomeronasal 2, receptor 69
Gene Aliases
-
Chromosome Location
Chr.7: 85406376 - 85415676 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105182.1 NP_001098652.1 3 - 3 545 158

View Full Product Details