Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Evpl
Entrez Gene ID
14027
Gene Name
envoplakin
Gene Aliases
-
Chromosome Location
Chr.11: 116220559 - 116238147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025276.3 NP_079552.3 3 - 4 445 56

View Full Product Details