Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mb21d2
- Entrez Gene ID
-
239796
- Gene Name
- Mab-21 domain containing 2
- Gene Aliases
-
1600021P15Rik, A430031N04, C87006
- Chromosome Location
-
Chr.16: 28826176 - 28929698 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK079722.1 |
- |
1 - 2 |
78 |
86 |
View Full Product Details