Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ostn
Entrez Gene ID
239790
Gene Name
osteocrin
Gene Aliases
Ostc
Chromosome Location
Chr.16: 27306983 - 27351209 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198112.2 NP_932780.1 2 - 3 160 63
GenBank mRNA AY395730.1 - 2 - 3 160 63
AY573932.1 - 2 - 3 312 63
BC117063.1 - 2 - 3 148 63
BC117089.1 - 2 - 3 148 63

View Full Product Details