Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rexo4
Entrez Gene ID
227656
Gene Name
REX4, RNA exonuclease 4 homolog (S. cerevisiae)
Gene Aliases
Gm111, Xpmc2h
Chromosome Location
Chr.2: 26953563 - 26964539 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK079733.1 - 1 - 1 1379 72

View Full Product Details