The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025984.2 | NP_080260.1 | 2 - 2 | 548 | 171 |
GenBank mRNA | AA755466.1 | - | 1 - 1 | 171 | |
AK008643.2 | - | 2 - 2 | 549 | 171 | |
AK009102.1 | - | 2 - 2 | 550 | 171 | |
BC108406.1 | - | 2 - 2 | 524 | 171 |