Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sval3
Entrez Gene ID
387564
Gene Name
seminal vesicle antigen-like 3
Gene Aliases
-
Chromosome Location
Chr.6: 41968140 - 41973090 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003952.1 NP_001003952.1 3 - 4 309 126
GenBank mRNA AB098509.1 - 3 - 4 309 126

View Full Product Details