Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
4930520M14Rik
Entrez Gene ID
78182
Gene Name
RIKEN cDNA 4930520M14 gene
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK019689.1 - 1 - 2 401 75


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715405.1 - 1 - 2 401 Chr. 5 - 139215495 - 139216356 [-] on Build GRCm38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream dynein, axonemal, assembly factor 5 Dnaaf5 BC053401, C76907, Heatr2 -29.0kb Anti-sense Chr. - 139150223 - 139186505 on Build GRCm38
Downstream golgi to ER traffic protein 4 homolog (S. cerevisiae) Get4 1110007L15Rik, AW412535, Cee 36.0kb Anti-sense Chr. - 139252324 - 139270050 on Build GRCm38

View Full Product Details