The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019866.1 | NP_063919.1 | 6 - 6 | 1583 | 85 |
GenBank mRNA | U87620.1 | - | 1 - 1 | 1057 | 84 |