Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Aasdhppt
- Entrez Gene ID
-
67618
- Gene Name
- aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
- Gene Aliases
-
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5
- Chromosome Location
-
Chr.9: 4294793 - 4309509 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK031766.1 |
- |
2 - 3 |
410 |
87 |
AK088977.1 |
- |
2 - 3 |
406 |
87 |
View Full Product Details