Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chchd2
Entrez Gene ID
14004
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Gene Aliases
AL033347, Etohi6
Chromosome Location
Chr.5: 129881161 - 129887470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC037682.1 - 2 - 2 1574 169
BC083126.1 - 2 - 2 1537 169

View Full Product Details