Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- Btg3
- Entrez Gene ID
-
12228
- Gene Name
- B cell translocation gene 3
- Gene Aliases
-
ANA, tob5
- Chromosome Location
-
Chr.16: 78359860 - 78377181 on Build GRCm38
- Gene Symbol
- Gm7334
- Entrez Gene ID
-
654432
- Gene Name
- B-cell translocation gene 3 pseudogene
- Gene Aliases
-
EG654432
- Chromosome Location
-
Chr.17: 50698525 - 50699892 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002700.1 |
|
1 - 1 |
648 |
148 |
View Full Product Details