The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198886.3 | NP_942589.3 | 2 - 2 | 1784 | 110 |
GenBank mRNA | BC020447.1 | - | 2 - 2 | 1723 | 110 |
BC062159.1 | - | 2 - 2 | 1784 | 110 |