The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011654.2 | NP_035784.1 | 4 - 4 | 1316 | 169 |
GenBank mRNA | AK075955.1 | - | 4 - 4 | 1363 | 169 |
AK137885.1 | - | 4 - 4 | 1360 | 169 | |
AK150128.1 | - | 4 - 4 | 1363 | 169 | |
AK150179.1 | - | 4 - 4 | 1363 | 169 | |
AK150968.1 | - | 4 - 4 | 1362 | 169 | |
AK151199.1 | - | 4 - 4 | 1360 | 169 | |
AK151809.1 | - | 4 - 4 | 1360 | 169 | |
AK152298.1 | - | 4 - 4 | 1360 | 169 | |
AK153064.1 | - | 4 - 4 | 1360 | 169 | |
AK153254.1 | - | 4 - 4 | 1360 | 169 | |
AK158958.1 | - | 4 - 4 | 1492 | 169 | |
AK164428.1 | - | 4 - 4 | 1363 | 169 | |
AK168770.1 | - | 4 - 4 | 1360 | 169 | |
AK169075.1 | - | 4 - 4 | 1360 | 169 | |
AK169092.1 | - | 4 - 4 | 1363 | 169 | |
AK169130.1 | - | 4 - 4 | 1363 | 169 | |
AK169665.1 | - | 4 - 4 | 1363 | 169 | |
AK191172.1 | - | 1 - 1 | 170 | 169 | |
AK194180.1 | - | 1 - 1 | 169 | ||
AK198269.1 | - | 1 - 1 | 170 | 169 | |
AK199828.1 | - | 1 - 1 | 117 | 169 | |
AK200029.1 | - | 1 - 1 | 141 | 169 | |
AK200084.1 | - | 1 - 1 | 170 | 169 | |
AK203041.1 | - | 1 - 1 | 93 | 169 | |
AK211470.1 | - | 1 - 1 | 138 | 169 | |
BC002219.1 | - | 4 - 4 | 1350 | 169 | |
BC004790.1 | - | 2 - 2 | 951 | 169 | |
BC008117.1 | - | 4 - 4 | 1343 | 169 | |
BC020390.1 | - | - | 1203 | 169 | |
BC063777.1 | - | 4 - 4 | 1313 | 169 | |
BC083120.1 | - | 4 - 4 | 1341 | 169 | |
BC098321.1 | - | 4 - 4 | 1343 | 169 | |
BC108337.1 | - | 4 - 4 | 1342 | 169 | |
BC108394.1 | - | 4 - 4 | 1316 | 169 | |
M13446.1 | - | 4 - 4 | 1340 | 169 | |
M28727.1 | - | 2 - 2 | 964 | 169 |