Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Amy2a2
- Entrez Gene ID
-
100043688
- Gene Name
- amylase 2a2
- Gene Aliases
-
ENSMUSG00000074265, OTTMUSG00000022431, mAmy2-5
- Chromosome Location
-
Chr.3: 113345049 - 113356658 on Build GRCm38
- Gene Symbol
- Amy2a3
- Entrez Gene ID
-
100043686
- Gene Name
- amylase 2a3
- Gene Aliases
-
OTTMUSG00000022430, mAmy2-4
- Chromosome Location
-
Chr.3: 113312449 - 113324052 on Build GRCm38
- Gene Symbol
- Amy2a4
- Entrez Gene ID
-
100043684
- Gene Name
- amylase 2a4
- Gene Aliases
-
ENSMUSG00000074267, OTTMUSG00000022433, mAmy2-3
- Chromosome Location
-
Chr.3: 113279837 - 113291449 on Build GRCm38
- Gene Symbol
- Amy2a5
- Entrez Gene ID
-
109959
- Gene Name
- amylase 2a5
- Gene Aliases
-
1810008N23Rik, Amy-2, Amy2, mAmy2-2
- Chromosome Location
-
Chr.3: 113249178 - 113258699 on Build GRCm38
View Full Product Details