The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031192.3 | NP_112469.1 | 2 - 3 | 277 | 141 |
GenBank mRNA | AK052685.1 | - | 2 - 3 | 277 | 141 |
AK085309.1 | - | 2 - 3 | 277 | 141 | |
BC061053.1 | - | 2 - 3 | 275 | 141 | |
X16642.1 | - | 2 - 3 | 259 | 141 |