The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009493.2 | NP_033519.1 | 1 - 2 | 302 | 178 |
GenBank mRNA | AF011414.1 | - | 1 - 2 | 302 | 178 |