Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cabs1
Entrez Gene ID
70977
Gene Name
calcium binding protein, spermatid specific 1
Gene Aliases
4931407G18Rik, AI428159, Clph
Chromosome Location
Chr.5: 87979451 - 87981541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027631.2 NP_081907.1 1 - 2 1338 122
GenBank mRNA AK076636.1 - 1 - 2 1339 122
BC049750.1 - 1 - 2 1338 122

View Full Product Details