The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004143.4 | NP_001004143.2 | 10 - 11 | 1493 | 138 |
GenBank mRNA | AK034492.1 | - | 10 - 11 | 1468 | 138 |
AK084022.1 | - | 6 - 7 | 651 | 138 | |
AK155071.1 | - | 10 - 11 | 1494 | 138 | |
AK160992.1 | - | 10 - 11 | 1402 | 138 | |
AK220368.1 | - | 10 - 11 | 1454 | 138 | |
BC026701.1 | - | 3 - 4 | 265 | 138 | |
BC052540.1 | - | 6 - 7 | 788 | 138 | |
BC058419.1 | - | 3 - 4 | 312 | 138 | |
BC080737.1 | - | 10 - 11 | 1364 | 138 |