Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqsec2
Entrez Gene ID
245666
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
Brag1, mKIAA0522
Chromosome Location
Chr.X: 152144183 - 152225237 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005475.2 NP_001005475.1 10 - 11 2460 129
GenBank mRNA AK220345.1 - 4 - 5 495 129
AY451401.1 - 10 - 11 3220 129
CB236789.1 - - 410 129

View Full Product Details