The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163387.1 | NP_001156859.1 | 3 - 4 | 1005 | 129 |
NM_138666.3 | NP_619607.2 | 5 - 6 | 2005 | 129 | |
GenBank mRNA | AK122433.1 | - | 5 - 6 | 2006 | 129 |