The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011687.2 | NP_001011687.1 | 1 - 1 | 513 | 70 |
GenBank mRNA | BC132614.1 | - | 1 - 1 | 513 | 70 |
BC132616.1 | - | 1 - 1 | 513 | 70 |