Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bex4
Entrez Gene ID
406217
Gene Name
brain expressed gene 4
Gene Aliases
-
Chromosome Location
Chr.X: 136139045 - 136140439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212457.2 NP_997622.1 3 - 3 354 84
GenBank mRNA AK010400.1 - 3 - 3 357 84
AY833558.1 - 3 - 3 354 84
BC090663.1 - 3 - 3 331 84
BC116226.2 - 3 - 3 258 84

View Full Product Details