Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclk3
Entrez Gene ID
245038
Gene Name
doublecortin-like kinase 3
Gene Aliases
BC056929, C730036H08, Dcamkl3
Chromosome Location
Chr.9: 111439081 - 111489611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172928.5 NP_766516.2 3 - 4 2248 73
GenBank mRNA AK050312.1 - 3 - 4 2216 73
AK135010.1 - 3 - 4 2237 73
BC056929.1 - 2 - 3 1916 73
DQ286388.1 - 3 - 4 2217 73

View Full Product Details