The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024414.2 | NP_077725.1 | 10 - 10 | 1396 | 92 |
GenBank mRNA | D45207.1 | - | - | 1392 | 92 |