The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012323.1 | NP_001012323.1 | 7 - 7 | 744 | 105 |
GenBank mRNA | BC089613.1 | - | 7 - 7 | 744 | 105 |
BC092096.1 | - | 7 - 7 | 770 | 105 |