Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptxr
Entrez Gene ID
73340
Gene Name
neuronal pentraxin receptor
Gene Aliases
1200009K17Rik, 1700036C17Rik, 5730406O18Rik, AI452369, AI785356, D15Bwg0580e, NPR, Npcd
Chromosome Location
Chr.15: 79786351 - 79804709 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030689.4 NP_109614.2 1 - 2 737 119
GenBank mRNA AK147546.1 - 1 - 2 794 119
AK147605.1 - 1 - 2 745 119
BC058962.1 - 1 - 2 736 119
BC098199.1 - 1 - 2 736 119

View Full Product Details