The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207000.2 | NP_996883.1 | 9 - 9 | 1821 | 137 |
GenBank mRNA | AF336305.1 | - | 9 - 9 | 1822 | 138 |
AK018355.1 | - | 9 - 9 | 1889 | 137 | |
AK032636.1 | - | 9 - 9 | 1905 | 137 |