The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033418.4 | NP_001028590.1 | 4 - 5 | 321 | 72 |
NM_001301682.1 | NP_001288611.1 | 3 - 4 | 291 | 72 | |
GenBank mRNA | AK136662.1 | - | 4 - 5 | 322 | 72 |
BC147454.1 | - | 4 - 5 | 306 | 72 | |
BC147459.1 | - | 4 - 5 | 306 | 72 | |
BC171987.1 | - | 3 - 4 | 276 | 72 | |
BC171991.1 | - | 3 - 4 | 276 | 72 | |
DQ227498.1 | - | 4 - 5 | 301 | 72 |