Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scoc
Entrez Gene ID
56367
Gene Name
short coiled-coil protein
Gene Aliases
0610011C06Rik, 2810002J11Rik, AA415030, SCOCO
Chromosome Location
Chr.8: 83434492 - 83458437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039137.3 NP_001034226.1 2 - 3 212 140
GenBank mRNA AK002521.1 - 2 - 3 166 140
BC147069.1 - 2 - 3 205 140

View Full Product Details