Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mettl7a2
- Entrez Gene ID
-
393082
- Gene Name
- methyltransferase like 7A2
- Gene Aliases
-
Gm921, UbiE2, Ubie
- Chromosome Location
-
Chr.15: 100353200 - 100361819 on Build GRCm38
- Gene Symbol
- Mettl7a3
- Entrez Gene ID
-
668178
- Gene Name
- methyltransferase like 7A3
- Gene Aliases
-
ENSMUSG00000058057, Gm10035, Ubie3
- Chromosome Location
-
Chr.15: 100334801 - 100347682 on Build GRCm38
View Full Product Details