The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008519.2 | NP_032545.1 | 2 - 2 | 1316 | 132 |
GenBank mRNA | AF044030.1 | - | 2 - 2 | 1312 | 132 |
AF110104.1 | - | 2 - 2 | 1263 | 132 | |
AK010895.1 | - | 2 - 2 | 1302 | 132 | |
AK155181.1 | - | 3 - 3 | 1109 | 132 | |
BC064063.1 | - | 2 - 2 | 1247 | 132 |