The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159652.1 | NP_001153124.1 | 1 - 1 | 1851 | 68 |
GenBank mRNA | AK005968.1 | - | 1 - 1 | 842 | 68 |
AK015060.1 | - | 1 - 1 | 829 | 68 | |
AK015270.1 | - | 1 - 1 | 838 | 68 | |
AK016613.1 | - | 1 - 1 | 829 | 68 |