The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039666.1 | NP_001034755.1 | 5 - 6 | 978 | 78 |
GenBank mRNA | BC107359.1 | - | 5 - 6 | 978 | 78 |
BC107360.2 | - | 5 - 6 | 976 | 78 | |
BC119521.2 | - | 3 - 4 | 738 | 78 | |
BC119522.2 | - | 5 - 6 | 978 | 78 |